Assay Details
| Gene Symbol: Btc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007568.5 | NP_031594.1 | 3 - 4 | 549 | 61 | |
| GenBank mRNA | AK046658.1 | - | 3 - 4 | 379 | 61 | |
| AK050206.1 | - | 3 - 4 | 365 | 61 | ||
| AK076272.1 | - | 3 - 4 | 469 | 61 | ||
| AK164633.1 | - | 3 - 4 | 550 | 61 | ||
| BC119152.1 | - | 3 - 4 | 415 | 8733901 | 61 | |
| BC119154.1 | - | 3 - 4 | 415 | 8733903 | 61 | |
| L08394.1 | - | 3 - 4 | 388 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Btc
Genomic Map