Assay Details
| Gene Symbol: Crhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007762.4 | NP_031788.1 | 4 - 5 | 521 | 84 | |
| GenBank mRNA | AB375515.1 | - | 4 - 5 | 331 | 84 | |
| AF369654.1 | - | 2 - 3 | 114 | 84 | ||
| AF483484.1 | - | 4 - 5 | 331 | 84 | ||
| AF483485.1 | - | 4 - 5 | 331 | 84 | ||
| AK144430.1 | - | 4 - 5 | 589 | 84 | ||
| AK158806.1 | - | 4 - 5 | 522 | 84 | ||
| BC103675.1 | - | 4 - 5 | 349 | 40046992 | 84 | |
| BC104735.1 | - | 4 - 5 | 349 | 40046995 | 84 | |
| BC105675.1 | - | 4 - 5 | 349 | 40046993 | 84 | |
| FJ668671.1 | - | 3 - 4 | 211 | 84 | ||
| X72305.1 | - | 4 - 5 | 362 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Crhr1
Genomic Map