Assay Details
| Gene Symbol: Crhr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007762.4 | NP_031788.1 | 6 - 7 | 742 | 96 | |
| GenBank mRNA | AB375515.1 | - | 6 - 7 | 552 | 96 | |
| AF483484.1 | - | 6 - 7 | 552 | 96 | ||
| AF483485.1 | - | 6 - 7 | 552 | 96 | ||
| AK144430.1 | - | 6 - 7 | 810 | 96 | ||
| AK158806.1 | - | 6 - 7 | 743 | 96 | ||
| BC103675.1 | - | 6 - 7 | 570 | 40046992 | 96 | |
| BC104735.1 | - | 7 - 8 | 570 | 40046995 | 96 | |
| BC105673.1 | - | 7 - 8 | 646 | 40046994 | 96 | |
| BC105675.1 | - | 6 - 7 | 570 | 40046993 | 96 | |
| FJ668671.1 | - | 5 - 6 | 432 | 96 | ||
| X72305.1 | - | 6 - 7 | 583 | 96 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC105673.1 transcripts.
Set Membership: |
|

Crhr1
Genomic Map