Assay Details
| Gene Symbol: Dhh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007857.4 | NP_031883.1 | 2 - 3 | 830 | 87 | |
| GenBank mRNA | AK032466.1 | - | 2 - 3 | 813 | 87 | |
| AK033392.1 | - | 2 - 3 | 831 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Effects of gestational diethylstilbestrol treatment on male and female gonads during early embryonic development.
- Authors
- Ikeda Y, Tanaka H, Esaki M
- Journal
- Endocrinology - Page No 3970 (Fri Aug 01 00:00:00 UTC 2008)
- Volume (Year)
- 149 (2008)
- Product Usage
- -

Dhh
Genomic Map