Assay Details
| Gene Symbol: F2rl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007975.3 | NP_032001.2 | 1 - 2 | 136 | 103 | |
| GenBank mRNA | AF080215.1 | - | 1 - 2 | 113 | 103 | |
| AK036427.1 | - | 1 - 2 | 137 | 103 | ||
| AK153720.1 | - | 1 - 2 | 130 | 103 | ||
| AK156729.1 | - | 1 - 2 | 137 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
G-protein coupled receptor signaling pathway
blood coagulation
hemostasis
positive regulation of release of sequestered calcium ion into cytosol
thrombin receptor signaling pathway
signal transducer activity
G-protein coupled receptor activity
thrombin receptor activity
signal transduction
G-protein coupled receptor signaling pathway
blood coagulation
hemostasis
positive regulation of release of sequestered calcium ion into cytosol
thrombin receptor signaling pathway
signal transducer activity
G-protein coupled receptor activity
thrombin receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011573 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | AK036427.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | AK153720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | AK156729.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | BC119461.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | AF080215.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | AK036427.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | AK153720.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | AK156729.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | BC119461.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P431636 | AF080215.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 859913 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165300 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | AK036427.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | AK153720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | AK156729.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | BC119461.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | AF080215.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | AK036427.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | AK153720.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | AK156729.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | BC119461.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P431636 | AF080215.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011573 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | AK036427.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | AK153720.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | AK156729.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | BC119461.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | AF080215.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | AK036427.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | AK153720.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | AK156729.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | BC119461.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P431636 | AF080215.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962808 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962826 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962802 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25962817 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165300 | - | - |
| Whole Mouse Genome G4122A | A_51_P431636 | AK036427.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | AK153720.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | AK156729.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | BC119461.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | AF080215.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | AK036427.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | AK153720.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | AK156729.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | BC119461.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P431636 | AF080215.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100317 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045350 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045365 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045345 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07200708 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25962858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962856 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06692086 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962835 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962818 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07045350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962828 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07012289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25962837 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572862 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572863 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421288_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572861 | - | - |
| Mouse Genome 430 2.0 Array | 1421288_at | - | - |
| Mouse Genome 430 2.0 Array | 1421288_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800006193.mm.1 | NM_007975.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006193.mm.1 | TC0800000835.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006194.mm.1 | NM_007975.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006194.mm.1 | TC0800000835.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800006191.mm.1 | NM_007975.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800006191.mm.1 | TC0800000835.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421288_PM_at | - | - |
| Mouse Expression Set 430 | 1421288_at | - | - |
| Murine Genome U74v2 Set | 94191_PM_at | - | - |
| Murine Genome U74v2 Set | 94191_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421288_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421288_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421288_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421288_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 367638 | - | - |
| ABI Mouse Genome Survey Microarray | 819057 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6620010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006620010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2737523 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2737523 | - | - |
| mouse-6 v1.1 (Illumina) | scl33668.2_157-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6620010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33668.2_157-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6620010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2737523 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2737523 | - | - |

F2rl3
Genomic Map