Assay Details
| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008072.2 | NP_032098.2 | 5 - 6 | 661 | 82 | |
| GenBank mRNA | AF326546.1 | - | 5 - 6 | 554 | 82 | |
| AF326564.1 | - | 5 - 6 | 554 | 82 | ||
| BC116832.1 | - | 5 - 6 | 596 | 40126151 | 82 | |
| BC116836.1 | - | 5 - 6 | 596 | 40126155 | 82 | |
| S42882.1 | - | 5 - 6 | 654 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabrd
Genomic Map