Assay Details
| Gene Symbol: Hspa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002012.1 | NP_001002012.1 | 2 | 1872 | 65 | |
| NM_008301.4 | NP_032327.2 | 1 | 1917 | 65 | ||
| GenBank mRNA | AK132261.1 | - | 1 | 1879 | 65 | |
| BC004714.1 | - | 1 | 1884 | 3499661 | 65 | |
| BC052350.1 | - | 2 | 1872 | 6772789 | 65 | |
More Information
Set Membership: |
|

Hspa2
Genomic Map