Assay Details
| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008350.4 | NP_032376.1 | 1 - 2 | 156 | 133 | |
| GenBank mRNA | U03421.1 | - | 1 - 2 | 49 | 133 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_008350.4,U03421.1 transcripts.
Set Membership: |
|

Il11
Genomic Map