Assay Details
| Gene Symbol: Il11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290423.1 | NP_001277352.1 | 4 - 5 | 483 | 130 | |
| NM_008350.4 | NP_032376.1 | 4 - 5 | 582 | 130 | ||
| GenBank mRNA | AK132897.1 | - | 4 - 5 | 484 | 130 | |
| BC134354.1 | - | 4 - 5 | 444 | 40054168 | 130 | |
| U03421.1 | - | 4 - 5 | 475 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell proliferation
positive regulation of peptidyl-serine phosphorylation
positive regulation of MAPK cascade
positive regulation of transcription from RNA polymerase II promoter
negative regulation of hormone secretion
positive regulation of peptidyl-tyrosine phosphorylation
cytokine activity
interleukin-11 receptor binding
growth factor activity
positive regulation of cell proliferation
positive regulation of peptidyl-serine phosphorylation
positive regulation of MAPK cascade
positive regulation of transcription from RNA polymerase II promoter
negative regulation of hormone secretion
positive regulation of peptidyl-tyrosine phosphorylation
cytokine activity
interleukin-11 receptor binding
growth factor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267783 | NM_008350.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267783 | AK132897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267783 | U03421.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267783 | NM_008350.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267783 | AK132897.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267783 | U03421.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513234 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666349 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 889934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 448996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 321890 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132874 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267783 | NM_008350.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267783 | AK132897.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267783 | U03421.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267783 | NM_008350.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267783 | AK132897.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267783 | U03421.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267783 | NM_008350.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267783 | AK132897.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267783 | U03421.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267783 | NM_008350.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267783 | AK132897.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P267783 | U03421.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950422 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06114492 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01475924 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132874 | - | - |
| Whole Mouse Genome G4122A | A_51_P267783 | NM_008350.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267783 | AK132897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267783 | U03421.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267783 | NM_008350.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267783 | AK132897.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267783 | U03421.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124809 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132874 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05899477 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05899486 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05899482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06577222 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06434621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06148333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06526302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05899479 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559670 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559668 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559669 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559671 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559672 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449982_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559667 | - | - |
| Mouse Genome 430 2.0 Array | 1449982_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449982_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16156_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16156_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019844.mm.1 | NM_008350.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019844.mm.1 | NM_008350.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019844.mm.1 | TC0700002171.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019843.mm.1 | NM_008350.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019843.mm.1 | NM_008350.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019843.mm.1 | TC0700002171.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019845.mm.1 | NM_008350.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019845.mm.1 | NM_008350.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019845.mm.1 | TC0700002171.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019846.mm.1 | NM_008350.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019846.mm.1 | NM_008350.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019846.mm.1 | TC0700002171.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449982_PM_at | - | - |
| Mouse Expression Set 430 | 1449982_at | - | - |
| Murine Genome U74v2 Set | 92266_at | - | - |
| Murine Genome U74v2 Set | 92266_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449982_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449982_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449982_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449982_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 487854 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1740398 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740398 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243862 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243862 | - | - |
| mouse-6 v1.1 (Illumina) | scl31801.4.1_81-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740398 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31801.4.1_81-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740398 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1243862 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243862 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31801.4.1_81-S | - | - |

Il11
Genomic Map