| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100033406.mm.1 |
NM_008379.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100033406.mm.1 |
NM_008379.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100033406.mm.1 |
TC1100003640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100033409.mm.1 |
NM_008379.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100033409.mm.1 |
NM_008379.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100033409.mm.1 |
TC1100003640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100033407.mm.1 |
NM_008379.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100033407.mm.1 |
NM_008379.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100033407.mm.1 |
TC1100003640.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100033408.mm.1 |
NM_008379.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100033408.mm.1 |
NM_008379.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100033408.mm.1 |
TC1100003640.mm.1 |
MTA TC |