Assay Details
| Gene Symbol: Mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008595.2 | NP_032621.1 | 6 - 7 | 988 | 136 | |
| GenBank mRNA | AF015769.1 | - | 6 - 7 | 815 | 136 | |
| AK143523.1 | - | 6 - 7 | 1022 | 136 | ||
| BC010983.1 | - | 6 - 7 | 1010 | 3980692 | 136 | |
| U94349.1 | - | 6 - 7 | 988 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfng
Genomic Map