Assay Details
| Gene Symbol: Cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008795.2 | NP_032821.1 | 1 - 2 | 279 | 115 | |
| GenBank mRNA | AK004998.1 | - | 1 - 2 | 223 | 115 | |
| AK171569.1 | - | 1 - 2 | 60 | 115 | ||
| X69026.1 | - | 1 - 2 | 296 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004998.1, NM_008795.2, X69026.1 transcripts.
Set Membership: |
|

Cdk18
Genomic Map