Assay Details
| Gene Symbol: Pfkl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008826.4 | NP_032852.2 | 20 - 21 | 2203 | 61 | |
| GenBank mRNA | AK030511.1 | - | 20 - 21 | 2158 | 61 | |
| AK030892.1 | - | 13 - 14 | 1302 | 61 | ||
| AK036318.1 | - | 20 - 21 | 2136 | 61 | ||
| AK081313.1 | - | 20 - 21 | 2154 | 61 | ||
| AK159328.1 | - | 20 - 21 | 2148 | 61 | ||
| AK168660.1 | - | 20 - 21 | 2159 | 61 | ||
| BC020097.1 | - | 20 - 21 | 2157 | 4014395 | 61 | |
| BC029336.1 | - | 20 - 21 | 2146 | 61 | ||
| J03928.1 | - | 20 - 21 | 2132 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pfkl
Genomic Map