Assay Details
| Gene Symbol: Pitx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008852.4 | NP_032878.1 | 1 - 2 | 126 | 97 | |
| GenBank mRNA | AF005772.1 | - | 1 - 2 | 122 | 97 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF005772.1,NM_008852.4 transcripts.
Set Membership: |
|

Pitx3
Genomic Map