Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100005742.mm.1 |
NM_009208.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005742.mm.1 |
NM_009208.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005742.mm.1 |
TC0100000665.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005740.mm.1 |
NM_009208.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005740.mm.1 |
NM_009208.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005740.mm.1 |
TC0100000665.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005734.mm.1 |
NM_009208.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005734.mm.1 |
NM_009208.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005734.mm.1 |
TC0100000665.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100005741.mm.1 |
NM_009208.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005741.mm.1 |
NM_009208.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100005741.mm.1 |
TC0100000665.mm.1 |
MTA TC |