Assay Details
| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009364.3 | NP_033390.1 | 2 - 3 | 304 | 99 | |
| GenBank mRNA | AK132268.1 | - | 2 - 3 | 326 | 99 | |
| BC021639.1 | - | 2 - 3 | 339 | 4023742 | 99 | |
| D50586.1 | - | 2 - 3 | 309 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216496 | BC021639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216496 | D50586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216496 | NM_009364.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216496 | AK132268.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216496 | BC021639.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216496 | D50586.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216496 | NM_009364.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P216496 | AK132268.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 343333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661417 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116269 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216496 | BC021639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216496 | D50586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216496 | NM_009364.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216496 | AK132268.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216496 | BC021639.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216496 | D50586.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216496 | NM_009364.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P216496 | AK132268.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216496 | BC021639.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216496 | D50586.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216496 | NM_009364.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216496 | AK132268.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216496 | BC021639.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216496 | D50586.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216496 | NM_009364.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216496 | AK132268.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201781 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201761 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201782 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201772 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201786 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24201757 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116269 | - | - |
| Whole Mouse Genome G4122A | A_51_P216496 | BC021639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216496 | D50586.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216496 | NM_009364.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216496 | AK132268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216496 | BC021639.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216496 | D50586.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216496 | NM_009364.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P216496 | AK132268.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119417 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116269 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05772318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05772321 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05772314 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05772313 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05772312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24201695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05772320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201743 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05772310 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05759368 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05772306 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201781 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05772322 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05260646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05388713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05525107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05112974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05772316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05546459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201778 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201786 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05546455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05772321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05772312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201725 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05257409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05257407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201755 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24201795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05388718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10542955 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542954 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542958 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542957 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10542956 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418547_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10542953 | - | - |
| Mouse Genome 430 2.0 Array | 1418547_at | - | - |
| Mouse Genome 430 2.0 Array | 1418547_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21789_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21789_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600014190.mm.1 | NM_009364.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014190.mm.1 | TC0600001793.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418547_at | - | - |
| Mouse Expression Set 430 | 1418547_PM_at | - | - |
| Murine Genome U74v2 Set | 94383_PM_at | - | - |
| Murine Genome U74v2 Set | 94383_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418547_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418547_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418547_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418547_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 384930 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3870324 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003870324 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2670204 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2670204 | - | - |
| mouse-6 v1.1 (Illumina) | scl29323.6.1_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3870324 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29323.6.1_14-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870324 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2670204 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2670204 | - | - |

Tfpi2
Genomic Map