Assay Details
| Gene Symbol: Tnfsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009404.3 | NP_033430.1 | 2 - 3 | 516 | 64 | |
| GenBank mRNA | AK155610.1 | - | 2 - 3 | 517 | 64 | |
| AK169471.1 | - | 2 - 3 | 489 | 64 | ||
| BC138767.1 | - | 2 - 3 | 495 | 8861789 | 64 | |
| BC138770.1 | - | 2 - 3 | 495 | 8861792 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
immune response
cell-cell signaling
positive regulation of interferon-gamma production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of activated T cell proliferation
regulation of cell proliferation
myeloid dendritic cell differentiation
positive regulation of cytotoxic T cell differentiation
receptor binding
cytokine activity
tumor necrosis factor receptor binding
tumor necrosis factor receptor superfamily binding
immune response
cell-cell signaling
positive regulation of interferon-gamma production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of activated T cell proliferation
regulation of cell proliferation
myeloid dendritic cell differentiation
positive regulation of cytotoxic T cell differentiation
receptor binding
cytokine activity
tumor necrosis factor receptor binding
tumor necrosis factor receptor superfamily binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31260063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31260061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31260062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31260065 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010038 | NM_009404.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2010038 | AK155610.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710794 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51552 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109852 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105561 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171382 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010038 | NM_009404.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2010038 | AK155610.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460004 | AK169471.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460004 | BC138767.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460004 | BC138770.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460004 | NM_009404.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P460004 | AK155610.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260082 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260065 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260081 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31260063 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109852 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105561 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171382 | - | - |
| Whole Mouse Genome G4122A | A_51_P460004 | AK169471.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460004 | BC138767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460004 | BC138770.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460004 | NM_009404.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P460004 | AK155610.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108255 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105561 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171382 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13545666 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13545663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13259233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13131864 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13259231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31260063 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31260062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31260065 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31260061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31260071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13330976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260038 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13330972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260060 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13321942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13309258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13309261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13131864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13126683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13545664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13294708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13471032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13440918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13511229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260101 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13126680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31260082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13321941 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10446230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446233 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422924_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10446229 | - | - |
| Mouse Genome 430 2.0 Array | 1422924_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422924_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21950_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21950_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700009887.mm.1 | NM_009404.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009887.mm.1 | NM_009404.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009887.mm.1 | TC1700001027.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700009888.mm.1 | NM_009404.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009888.mm.1 | NM_009404.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700009888.mm.1 | TC1700001027.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422924_PM_at | - | - |
| Mouse Expression Set 430 | 1422924_at | - | - |
| Murine Genome U74v2 Set | 92415_at | - | - |
| Murine Genome U74v2 Set | 92415_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422924_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422924_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422924_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422924_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 571554 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1190408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001190408 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2449157 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2449157 | - | - |
| mouse-6 v1.1 (Illumina) | scl50578.1.100_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1190408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50578.1.100_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1190408 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2449157 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2449157 | - | - |

Tnfsf9
Genomic Map