Assay Details
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK011710.1 | - | 3 - 4 | 240 | 78 | |
AK146363.1 | - | 3 - 4 | 250 | 78 | ||
AK164321.1 | - | 3 - 4 | 216 | 78 | ||
AK165998.1 | - | 3 - 4 | 227 | 78 | ||
AK168842.1 | - | 3 - 4 | 248 | 78 | ||
BC002022.1 | - | 3 - 4 | 236 | 3491126 | 78 | |
BY070840.1 | - | 3 - 4 | 276 | 78 | ||
Y13087.1 | - | 2 - 3 | 204 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
