Assay Details
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK011710.1 | - | 4 - 5 | 321 | 90 | |
AK146363.1 | - | 4 - 5 | 331 | 90 | ||
AK164321.1 | - | 4 - 5 | 297 | 90 | ||
AK165998.1 | - | 4 - 5 | 308 | 90 | ||
AK168842.1 | - | 4 - 5 | 329 | 90 | ||
BC002022.1 | - | 4 - 5 | 317 | 3491126 | 90 | |
Y13087.1 | - | 3 - 4 | 285 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22522750 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22522753 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22522752 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22522751 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | AW556149.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | BC002022.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | Y13087.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | AK011710.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | AK146363.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | AW556149.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | BC002022.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | Y13087.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | AK011710.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P215922 | AK146363.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473173 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926277 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724476 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496340 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298100 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180816 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113893 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168219 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | AW556149.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | BC002022.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | Y13087.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | AK011710.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | AK146363.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | AW556149.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | BC002022.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | Y13087.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | AK011710.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P215922 | AK146363.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | AW556149.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | BC002022.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | Y13087.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | AK011710.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | AK146363.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | AW556149.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | BC002022.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | Y13087.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | AK011710.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P215922 | AK146363.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522769 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522724 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522728 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22522763 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113893 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05071573 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168219 | - | - |
Whole Mouse Genome G4122A | A_51_P215922 | AW556149.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | BC002022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | Y13087.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | AK011710.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | AK146363.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | AW556149.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | BC002022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | Y13087.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | AK011710.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P215922 | AK146363.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102740 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168219 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113893 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02905997 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02905985 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02951859 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22522750 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22522752 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22522751 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22522753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22522768 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02534471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02677975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02966824 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02905987 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02813692 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02534475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522723 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522784 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02905992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02712549 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02677984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02905996 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522742 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02534476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02813695 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02534478 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522765 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02813693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02412426 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02601093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522724 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522780 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22522748 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02905997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02813694 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10496026 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496030 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496029 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496028 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496027 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496024 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496025 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10496031 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1415995_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1458715_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10496023 | - | - |
Mouse Genome 430 2.0 Array | 1415995_at | - | - |
Mouse Genome 430 2.0 Array | 1458715_at | - | - |
Mouse Genome 430 2.0 Array | 1415995_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1458715_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12368_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12368_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300010625.mm.1 | NM_009811.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300010625.mm.1 | TC0300001315.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300010622.mm.1 | NM_009811.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300010622.mm.1 | TC0300001315.mm.1 | MTA TC |
Mouse Expression Set 430 | 1415995_PM_at | - | - |
Mouse Expression Set 430 | 1415995_at | - | - |
Mouse Expression Set 430 | 1458715_at | - | - |
Mouse Expression Set 430 | 1458715_PM_at | - | - |
Murine Genome U74v2 Set | 94458_at | - | - |
Murine Genome U74v2 Set | 94458_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1415995_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1415995_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1415995_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1415995_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458715_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458715_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 804355 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5690239 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6420551 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006420551 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005690239 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2711604 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242974 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2711604 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1242974 | - | - |
mouse-6 v1.1 (Illumina) | scl0002068.1_1-S | - | - |
mouse-6 v1.1 (Illumina) | scl22623.8.1_50-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6420551 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5690239 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl22623.8.1_50-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002068.1_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690239 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420551 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2923864 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2923865 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2923864 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2923865 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0002068.1_1-S | - | - |
