Assay Details
| Gene Symbol: Cox7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009944.3 | NP_034074.1 | 1 - 2 | 73 | 78 | |
| GenBank mRNA | AF037370.1 | - | 1 - 2 | 36 | 78 | |
| AK079950.1 | - | 1 - 2 | 74 | 78 | ||
| BC060974.1 | - | 1 - 2 | 51 | 6543330 | 78 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF037370.1,AK079950.1,BC060974.1,NM_009944.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P148612 | NM_009944.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P148612 | AF037370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P148612 | AK079950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P148612 | BC060974.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P148612 | NM_009944.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P148612 | AF037370.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P148612 | AK079950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P148612 | BC060974.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191802 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P148612 | NM_009944.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P148612 | AF037370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P148612 | AK079950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P148612 | BC060974.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P148612 | NM_009944.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P148612 | AF037370.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P148612 | AK079950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P148612 | BC060974.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P148612 | NM_009944.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P148612 | AF037370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P148612 | AK079950.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P148612 | BC060974.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P148612 | NM_009944.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P148612 | AF037370.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P148612 | AK079950.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P148612 | BC060974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P148612 | NM_009944.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P148612 | AF037370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P148612 | AK079950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P148612 | BC060974.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P148612 | NM_009944.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P148612 | AF037370.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P148612 | AK079950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P148612 | BC060974.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25030316 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06041040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06502625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05942015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25030273 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10551838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551837 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418709_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551836 | - | - |
| Mouse Genome 430 2.0 Array | 1418709_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418709_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12865_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12865_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700004713.mm.1 | NM_009944.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700004713.mm.1 | NM_009944.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700004713.mm.1 | TC0700000552.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700004714.mm.1 | NM_009944.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700004714.mm.1 | NM_009944.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700004714.mm.1 | TC0700000552.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418709_PM_at | - | - |
| Mouse Expression Set 430 | 1418709_at | - | - |
| Murine Genome U74v2 Set | 102749_at | - | - |
| Murine Genome U74v2 Set | 102749_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418709_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418709_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418709_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418709_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 304467 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6770427 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6770427 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006770427 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240857 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240857 | - | - |
| mouse-6 v1.1 (Illumina) | scl0012865.1_86-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6770427 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012865.1_86-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770427 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240857 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240857 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0012865.1_86-S | - | - |

Cox7a1
Genomic Map