Assay Details
| Gene Symbol: Cox7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009944.3 | NP_034074.1 | 2 - 3 | 163 | 64 | |
| GenBank mRNA | AF037370.1 | - | 2 - 3 | 126 | 64 | |
| AK079950.1 | - | 2 - 3 | 164 | 64 | ||
| BC060974.1 | - | 2 - 3 | 141 | 6543330 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cox7a1
Genomic Map