Assay Details
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276387.1 | NP_001263316.1 | 6 - 7 | 1454 | 72 | |
| NM_010095.6 | NP_034225.1 | 6 - 7 | 1449 | 72 | ||
| GenBank mRNA | AK053013.1 | - | 6 - 7 | 1450 | 72 | |
| AK139262.1 | - | 6 - 7 | 1161 | 72 | ||
| BC049188.1 | - | 5 - 6 | 748 | 6312260 | 72 | |
| BC050922.1 | - | 5 - 6 | 971 | 6409737 | 72 | |
| U71189.1 | - | 5 - 6 | 634 | 72 | ||
| U82441.1 | - | 5 - 6 | 503 | 72 | ||
| U92703.1 | - | 5 - 6 | 854 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ebf2
Genomic Map