Assay Details
| Gene Symbol: Gabrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010252.4 | NP_034382.2 | 6 - 7 | 1161 | 103 | |
| GenBank mRNA | AF090375.1 | - | 1 - 2 | 99 | 103 | |
| AK032128.1 | - | 6 - 7 | 846 | 103 | ||
| AK034085.1 | - | 6 - 7 | 749 | 103 | ||
| AK034332.1 | - | 6 - 7 | 1162 | 103 | ||
| AK038468.1 | - | 6 - 7 | 898 | 103 | ||
| AK039068.1 | - | 6 - 7 | 859 | 103 | ||
| AK047752.1 | - | 6 - 7 | 911 | 103 | ||
| AK049956.1 | - | 6 - 7 | 899 | 103 | ||
| AK140399.1 | - | 6 - 7 | 911 | 103 | ||
| AK162884.1 | - | 7 - 8 | 844 | 103 | ||
| AK163541.1 | - | 6 - 7 | 869 | 104 | ||
| BC099939.1 | - | 6 - 7 | 870 | 30090762 | 103 | |
| X55272.1 | - | 6 - 7 | 944 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabrg1
Genomic Map