| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600016624.mm.1 |
NM_001136065.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016624.mm.1 |
NM_010433.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016624.mm.1 |
NM_010433.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016624.mm.1 |
TC0600002109.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016625.mm.1 |
NM_001136065.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016625.mm.1 |
NM_010433.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016625.mm.1 |
NM_010433.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016625.mm.1 |
TC0600002109.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600016627.mm.1 |
NM_001136065.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016627.mm.1 |
NM_010433.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016627.mm.1 |
NM_010433.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600016627.mm.1 |
TC0600002109.mm.1 |
MTA TC |