Assay Details
| Gene Symbol: Hipk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136065.2 | NP_001129537.1 | 3 - 4 | 1530 | 101 | |
| NM_001294143.1 | NP_001281072.1 | 3 - 4 | 1606 | 101 | ||
| NM_001294144.1 | NP_001281073.1 | 3 - 4 | 1606 | 101 | ||
| NM_010433.2 | NP_034563.2 | 3 - 4 | 1606 | 101 | ||
| GenBank mRNA | AF077659.1 | - | 3 - 4 | 1369 | 101 | |
| AF170301.1 | - | 3 - 4 | 1356 | 101 | ||
| AF170302.1 | - | 3 - 4 | 1356 | 101 | ||
| AF208292.1 | - | 3 - 4 | 1443 | 101 | ||
| AF273680.1 | - | 3 - 4 | 1327 | 101 | ||
| AF333791.1 | - | 3 - 4 | 1233 | 101 | ||
| AF333792.1 | - | 3 - 4 | 1233 | 101 | ||
| BC031904.1 | - | 3 - 4 | 1339 | 5368577 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hipk2
Genomic Map