Assay Details
| Gene Symbol: Hipk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136065.2 | NP_001129537.1 | 6 - 7 | 1915 | 76 | |
| NM_001294143.1 | NP_001281072.1 | 6 - 7 | 1991 | 76 | ||
| NM_001294144.1 | NP_001281073.1 | 6 - 7 | 1991 | 76 | ||
| NM_010433.2 | NP_034563.2 | 6 - 7 | 1991 | 76 | ||
| GenBank mRNA | AF077659.1 | - | 6 - 7 | 1754 | 76 | |
| AF170301.1 | - | 6 - 7 | 1741 | 76 | ||
| AF170302.1 | - | 6 - 7 | 1741 | 76 | ||
| AF208292.1 | - | 6 - 7 | 1828 | 76 | ||
| AF273680.1 | - | 6 - 7 | 1712 | 76 | ||
| AF333791.1 | - | 6 - 7 | 1618 | 76 | ||
| AF333792.1 | - | 6 - 7 | 1618 | 76 | ||
| AK019821.1 | - | 2 - 3 | 207 | 76 | ||
| BC031904.1 | - | 6 - 7 | 1724 | 5368577 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hipk2
Genomic Map