Assay Details
| Gene Symbol: Ifnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010510.1 | NP_034640.1 | 1 | 51 | 61 | |
| GenBank mRNA | BC119395.1 | - | 1 | 51 | 8734144 | 61 |
| BC119397.1 | - | 1 | 51 | 8734146 | 61 | |
| K00020.1 | - | 1 | 51 | 61 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC119395.1,BC119397.1,K00020.1,NM_010510.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (11)
Autophagy within the antigen donor cell facilitates efficient antigen cross-priming of virus-specific CD8+ T cells.
- Authors
- Uhl M, Kepp O, Jusforgues-Saklani H, Vicencio JM, Kroemer G, Albert ML
- Journal
- Cell Death Differ - Page No 991 (Wed Jul 01 00:00:00 UTC 2009)
- Volume (Year)
- 16 (2009)
- Product Usage
- -
Gender-dependent expression of murine Irf5 gene: implications for sex bias in autoimmunity.
- Authors
- Shen H, Panchanathan R, Rajavelu P, Duan X, Gould KA, Choubey D
- Journal
- J Mol Cell Biol - Page No 284 (Fri Oct 01 00:00:00 UTC 2010)
- Volume (Year)
- 2 (2010)
- Product Usage
- -
The alpha/beta interferon receptor provides protection against influenza virus replication but is dispensable for inflammatory response signaling.
- Authors
- Goodman AG, Zeng H, Proll SC, Peng X, Cilloniz C, Carter VS, Korth MJ, Tumpey TM, Katze MG
- Journal
- J Virol - Page No 2027 (Mon Feb 01 00:00:00 UTC 2010)
- Volume (Year)
- 84 (2010)
- Product Usage
- -
Type I interferons produced by resident renal cells may promote end-organ disease in autoantibody-mediated glomerulonephritis.
- Authors
- Fairhurst AM, Xie C, Fu Y, Wang A, Boudreaux C, Zhou XJ, Cibotti R, Coyle A, Connolly JE, Wakeland EK, Mohan C
- Journal
- J Immunol - Page No 6831 (Sun Nov 15 00:00:00 UTC 2009)
- Volume (Year)
- 183 (2009)
- Product Usage
- -
Bacterial lipopeptide triggers massive albuminuria in murine lupus nephritis by activating Toll-like receptor 2 at the glomerular filtration barrier.
- Authors
- Pawar RD, Castrezana-Lopez L, Allam R, Kulkarni OP, Segerer S, Radomska E, Meyer TN, Schwesinger CM, Akis N, Grone HJ, Anders HJ
- Journal
- Immunology - Page No e206 (Tue Sep 01 00:00:00 UTC 2009)
- Volume (Year)
- 128 (2009)
- Product Usage
- -
Identification of loss of function mutations in human genes encoding RIG-I and MDA5: implications for resistance to type I diabetes.
- Authors
- Shigemoto T, Kageyama M, Hirai R, Zheng J, Yoneyama M, Fujita T
- Journal
- J Biol Chem - Page No 13348 (Fri May 15 00:00:00 UTC 2009)
- Volume (Year)
- 284 (2009)
- Product Usage
- -
Tumor necrosis factor-alpha: alternative role as an inhibitor of osteoclast formation in vitro.
- Authors
- Balga R, Wetterwald A, Portenier J, Dolder S, Mueller C, Hofstetter W
- Journal
- Bone - Page No 325 (Tue Aug 01 00:00:00 UTC 2006)
- Volume (Year)
- 39 (2006)
- Product Usage
- -
Distinct RIG-I and MDA5 signaling by RNA viruses in innate immunity.
- Authors
- Loo YM, Fornek J, Crochet N, Bajwa G, Perwitasari O, Martinez-Sobrido L, Akira S, Gill MA, Garcia-Sastre A, Katze MG, Gale M Jr
- Journal
- J Virol - Page No 335 (Tue Jan 01 00:00:00 UTC 2008)
- Volume (Year)
- 82 (2008)
- Product Usage
- -
Tir8/Sigirr prevents murine lupus by suppressing the immunostimulatory effects of lupus autoantigens.
- Authors
- Lech M, Kulkarni OP, Pfeiffer S, Savarese E, Krug A, Garlanda C, Mantovani A, Anders HJ
- Journal
- J Exp Med - Page No 1879 (Mon Aug 04 00:00:00 UTC 2008)
- Volume (Year)
- 205 (2008)
- Product Usage
- -
Inflammatory responses to pneumovirus infection in IFN-alpha beta R gene-deleted mice.
- Authors
- Garvey TL, Dyer KD, Ellis JA, Bonville CA, Foster B, Prussin C, Easton AJ, Domachowske JB, Rosenberg HF
- Journal
- J Immunol - Page No 4735 (Sat Oct 01 00:00:00 UTC 2005)
- Volume (Year)
- 175 (2005)
- Product Usage
- -
Differential roles of estrogen receptors alpha and beta in control of B cell maturation and selection(1).
- Authors
- Hill L, Venkatesh J, Chinnasamy P, Grimaldi C, Diamond B
- Journal
- Mol Med - Page No (Mon Nov 22 00:00:00 UTC 2010)
- Volume (Year)
- (2010)
- Product Usage
- -

Ifnb1
Genomic Map