| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100015378.mm.1 |
NM_010892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015378.mm.1 |
NM_010892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015378.mm.1 |
TC0100001858.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100015381.mm.1 |
NM_010892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015381.mm.1 |
NM_010892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015381.mm.1 |
TC0100001858.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100015379.mm.1 |
NM_010892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015379.mm.1 |
NM_010892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015379.mm.1 |
TC0100001858.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100015374.mm.1 |
NM_010892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015374.mm.1 |
NM_010892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015374.mm.1 |
TC0100001858.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100015380.mm.1 |
NM_010892.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015380.mm.1 |
NM_010892.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015380.mm.1 |
TC0100001858.mm.1 |
MTA TC |