Assay Details
| Gene Symbol: Nos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010927.3 | NP_035057.1 | 10 - 11 | 1348 | 57 | |
| GenBank mRNA | AF065919.1 | - | 10 - 11 | 1345 | 57 | |
| AF065920.1 | - | 10 - 11 | 1345 | 57 | ||
| AF065921.2 | - | 10 - 11 | 1345 | 57 | ||
| AF065922.2 | - | 9 - 10 | 1218 | 57 | ||
| AF065923.2 | - | 9 - 10 | 1218 | 57 | ||
| AY090567.1 | - | 9 - 10 | 1168 | 57 | ||
| BC062378.1 | - | 10 - 11 | 1357 | 4978648 | 57 | |
| M84373.1 | - | 10 - 11 | 1349 | 57 | ||
| M87039.1 | - | 10 - 11 | 1420 | 57 | ||
| M92649.1 | - | 10 - 11 | 1356 | 57 | ||
| U43428.1 | - | 10 - 11 | 1236 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nos2
Genomic Map