Assay Details
| Gene Symbol: Nos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010927.3 | NP_035057.1 | 19 - 20 | 2413 | 87 | |
| GenBank mRNA | AF065919.1 | - | 19 - 20 | 2410 | 87 | |
| AF065920.1 | - | 19 - 20 | 2410 | 87 | ||
| AF065921.2 | - | 19 - 20 | 2410 | 87 | ||
| AF065922.2 | - | 18 - 19 | 2283 | 87 | ||
| AF065923.2 | - | 18 - 19 | 2283 | 87 | ||
| AY090567.1 | - | 18 - 19 | 2233 | 87 | ||
| BC062378.1 | - | 19 - 20 | 2422 | 4978648 | 87 | |
| M84373.1 | - | 19 - 20 | 2414 | 87 | ||
| M87039.1 | - | 19 - 20 | 2485 | 87 | ||
| M92649.1 | - | 19 - 20 | 2421 | 87 | ||
| U43428.1 | - | 19 - 20 | 2301 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nos2
Genomic Map