Assay Details
| Gene Symbol: Frzb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011356.4 | NP_035486.1 | 4 - 5 | 1356 | 140 | |
| GenBank mRNA | AK019093.2 | - | 4 - 5 | 1027 | 140 | |
| AK029941.1 | - | 4 - 5 | 1358 | 140 | ||
| BC016884.1 | - | 4 - 5 | 1203 | 4237554 | 140 | |
| U68058.1 | - | 4 - 5 | 812 | 140 | ||
| U88568.1 | - | 4 - 5 | 1164 | 140 | ||
| U91905.1 | - | 4 - 5 | 1598 | 140 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Frzb
Genomic Map