Assay Details
| Gene Symbol: Slc34a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011392.2 | NP_035522.2 | 2 - 3 | 216 | 95 | |
| GenBank mRNA | AB163921.1 | - | 221 | 95 | ||
| AB163922.1 | - | 2 - 3 | 160 | 95 | ||
| AK018740.2 | - | 2 - 3 | 171 | 95 | ||
| BC020529.1 | - | 2 - 3 | 159 | 4223484 | 95 | |
| CB952922.1 | - | 2 - 3 | 219 | 95 | ||
| L33878.1 | - | 1 - 2 | 143 | 95 | ||
| U22465.1 | - | 2 - 3 | 160 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc34a1
Genomic Map