Assay Details
| Gene Symbol: Slc34a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011392.2 | NP_035522.2 | 11 - 12 | 1395 | 59 | |
| GenBank mRNA | AB163921.1 | - | 1400 | 59 | ||
| AB163922.1 | - | 11 - 12 | 1339 | 59 | ||
| AK018740.2 | - | 11 - 12 | 1350 | 59 | ||
| AK032618.1 | - | 4 - 5 | 486 | 59 | ||
| BC020529.1 | - | 11 - 12 | 1338 | 4223484 | 59 | |
| L33878.1 | - | 10 - 11 | 1322 | 59 | ||
| U22465.1 | - | 11 - 12 | 1339 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc34a1
Genomic Map