Assay Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011485.4 | NP_035615.2 | 1 - 2 | 127 | 81 | |
| GenBank mRNA | AK019725.2 | - | 1 - 2 | 167 | 81 | |
| AK054470.1 | - | 1 - 2 | 127 | 81 | ||
| AK136327.1 | - | 1 - 2 | 137 | 81 | ||
| BC060970.1 | - | 1 - 2 | 138 | 81 | ||
| BC082283.1 | - | 1 - 2 | 138 | 30436512 | 81 | |
| L36062.1 | - | 1 - 2 | 143 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Star
Genomic Map