Assay Details
| Gene Symbol: Star | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011485.4 | NP_035615.2 | 2 - 3 | 232 | 82 | |
| GenBank mRNA | AK019725.2 | - | 2 - 3 | 272 | 82 | |
| AK054470.1 | - | 2 - 3 | 232 | 82 | ||
| AK136327.1 | - | 2 - 3 | 242 | 82 | ||
| BC060970.1 | - | 2 - 3 | 243 | 82 | ||
| BC082283.1 | - | 2 - 3 | 243 | 30436512 | 82 | |
| L36062.1 | - | 2 - 3 | 248 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Star
Genomic Map