Assay Details
| Gene Symbol: Ahsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013465.2 | NP_038493.1 | 2 - 3 | 429 | 80 | |
| GenBank mRNA | AK133523.1 | - | 2 - 3 | 400 | 80 | |
| AK149474.1 | - | 2 - 3 | 402 | 80 | ||
| AK167709.1 | - | 2 - 3 | 402 | 80 | ||
| BC012678.1 | - | 2 - 3 | 401 | 4193595 | 80 | |
| BC019822.1 | - | 2 - 3 | 360 | 5123689 | 80 | |
| S96534.1 | - | 2 - 3 | 385 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ahsg
Genomic Map