Assay Details
| Gene Symbol: Ahsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276450.1 | NP_001263379.1 | 3 - 4 | 491 | 74 | |
| NM_013465.2 | NP_038493.1 | 3 - 4 | 512 | 74 | ||
| GenBank mRNA | AK133523.1 | - | 3 - 4 | 483 | 74 | |
| AK149474.1 | - | 3 - 4 | 485 | 74 | ||
| AK149479.1 | - | 3 - 4 | 485 | 74 | ||
| AK167709.1 | - | 3 - 4 | 485 | 74 | ||
| BC012678.1 | - | 3 - 4 | 484 | 4193595 | 74 | |
| BC019822.1 | - | 3 - 4 | 443 | 5123689 | 74 | |
| S96534.1 | - | 3 - 4 | 468 | 74 | ||
More Information
Set Membership: |
|

Ahsg
Genomic Map