Assay Details
| Gene Symbol: Loxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013586.4 | NP_038614.2 | 1 - 2 | 67 | 104 | |
| GenBank mRNA | AF053368.1 | - | 1 - 2 | 98 | 104 | |
| AK030548.1 | - | 1 - 2 | 68 | 104 | ||
| AK145189.1 | - | 1 - 2 | 87 | 104 | ||
| BC011298.1 | - | 1 - 2 | 61 | 3488978 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF053368.1, AK030548.1, AK145189.1, BC011298.1, NM_013586.4 transcripts.
Set Membership: |
|

Loxl3
Genomic Map