Assay Details
| Gene Symbol: Loxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013586.4 | NP_038614.2 | 7 - 8 | 1350 | 79 | |
| GenBank mRNA | AF053368.1 | - | 7 - 8 | 1381 | 79 | |
| AK030548.1 | - | 7 - 8 | 1351 | 79 | ||
| BC011298.1 | - | 7 - 8 | 1344 | 3488978 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Loxl3
Genomic Map