Assay Details
| Gene Symbol: Loxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013586.4 | NP_038614.2 | 11 - 12 | 2039 | 82 | |
| GenBank mRNA | AF053368.1 | - | 11 - 12 | 2070 | 82 | |
| AK030548.1 | - | 11 - 12 | 2040 | 82 | ||
| BC011298.1 | - | 11 - 12 | 2033 | 3488978 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Loxl3
Genomic Map