| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500013977.mm.1 |
NM_001164047.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013977.mm.1 |
NM_001164047.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013977.mm.1 |
TC1500001798.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500013978.mm.1 |
NM_001164047.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013978.mm.1 |
NM_001164047.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013978.mm.1 |
NM_001164048.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013978.mm.1 |
NM_001164048.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013978.mm.1 |
TC1500001798.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500013975.mm.1 |
NM_001164047.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013975.mm.1 |
NM_001164047.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013975.mm.1 |
NM_001164048.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013975.mm.1 |
NM_001164048.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013975.mm.1 |
NM_013593.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013975.mm.1 |
NM_013593.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013975.mm.1 |
TC1500001798.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500013979.mm.1 |
NM_001164047.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013979.mm.1 |
NM_001164047.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013979.mm.1 |
TC1500001798.mm.1 |
MTA TC |