Assay Details
| Gene Symbol: Mylpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016754.5 | NP_058034.1 | 6 - 7 | 557 | 54 | |
| GenBank mRNA | AK003149.1 | - | 6 - 7 | 462 | 54 | |
| AK010483.1 | - | 6 - 7 | 558 | 54 | ||
| BC055869.1 | - | 6 - 7 | 441 | 4018694 | 54 | |
| U77943.1 | - | 5 - 6 | 404 | 54 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2170381 | NM_016754.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 698570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 788702 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141846 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124949 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2170381 | NM_016754.5 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504597 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25504586 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124949 | - | - |
| Whole Mouse Genome G4122A | A_52_P383572 | AK003149.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P383572 | AK010483.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P383572 | NM_016754.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P383572 | BC055869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P383572 | U77943.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112445 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138891 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124949 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06484465 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06484462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25504640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504580 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06484459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504595 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06484465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06525852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06056523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06484454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25504605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06525854 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10557582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557576 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557578 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557583 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557579 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557577 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557584 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10557580 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448371_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10557575 | - | - |
| Mouse Genome 430 2.0 Array | 1448371_at | - | - |
| Mouse Genome 430 2.0 Array | 1448371_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17907_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17907_PM_at | - | - |
| Mouse Expression Set 430 | 1448371_at | - | - |
| Mouse Expression Set 430 | 1448371_PM_at | - | - |
| Murine Genome U74v2 Set | 162101_PM_f_at | - | - |
| Murine Genome U74v2 Set | 162101_f_at | - | - |
| Murine Genome U74v2 Set | 92881_PM_at | - | - |
| Murine Genome U74v2 Set | 92881_at | - | - |
| Murine Genome U74v2 Set | 170347_f_at | - | - |
| Murine Genome U74v2 Set | 170347_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448371_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448371_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448371_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448371_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 926434 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6110092 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110092 | - | - |
| mouse-6 v1.1 (Illumina) | scl32040.11.1_26-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110092 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32040.11.1_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110092 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2977331 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2977331 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2745496 | - | - |

Mylpf
Genomic Map