Assay Details
| Gene Symbol: Apba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018758.2 | NP_061228.1 | 5 - 6 | 1106 | 61 | |
| GenBank mRNA | AF070975.1 | - | 5 - 6 | 1060 | 61 | |
| AK045965.1 | - | 5 - 6 | 1107 | 61 | ||
| AK082046.1 | - | 1026 | 61 | |||
| AK169462.1 | - | 5 - 6 | 1090 | 61 | ||
| AK170182.1 | - | 5 - 6 | 1054 | 61 | ||
| BC005423.1 | - | 5 - 6 | 1038 | 3487448 | 61 | |
| BC009666.1 | - | 5 - 6 | 1050 | 3490710 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apba3
Genomic Map