Assay Details
| Gene Symbol: Apba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018758.2 | NP_061228.1 | 10 - 11 | 1913 | 70 | |
| GenBank mRNA | AF070975.1 | - | 10 - 11 | 1867 | 70 | |
| AK045965.1 | - | 10 - 11 | 1914 | 70 | ||
| AK169462.1 | - | 10 - 11 | 1897 | 70 | ||
| AK170182.1 | - | 10 - 11 | 1861 | 70 | ||
| BC005423.1 | - | 10 - 11 | 1845 | 3487448 | 70 | |
| BC009666.1 | - | 10 - 11 | 1857 | 3490710 | 70 | |
| BQ031518.1 | - | 1 - 2 | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977538 | BC009666.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977538 | BQ031518.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977538 | AK169462.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977538 | AK170182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977538 | BC005423.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980981 | BC009666.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980981 | BQ031518.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980981 | AK169462.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980981 | AK170182.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980981 | BC005423.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506897 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866057 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888972 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134012 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977538 | BC009666.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977538 | BQ031518.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977538 | AK169462.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977538 | AK170182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977538 | BC005423.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980981 | BC009666.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980981 | BQ031518.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980981 | AK169462.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980981 | AK170182.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980981 | BC005423.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288474 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27288498 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134012 | - | - |
| Whole Mouse Genome G4122A | A_51_P285960 | BC009666.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P285960 | BQ031518.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P285960 | AK169462.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P285960 | AK170182.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P285960 | BC005423.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134012 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08624232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27288397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08624242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08624238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08624235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08624243 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27288468 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10365060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365065 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365068 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365057 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365067 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365066 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10365063 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416182_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10365056 | - | - |
| Mouse Genome 430 2.0 Array | 1454720_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1416182_at | - | - |
| Mouse Genome 430 2.0 Array | 1454720_at | - | - |
| Mouse Genome 430 2.0 Array | 1416182_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57267_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 57267_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000006986.mm.1 | NM_018758.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006986.mm.1 | NM_018758.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000006986.mm.1 | TC1000000891.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416182_at | - | - |
| Mouse Expression Set 430 | 1416182_PM_at | - | - |
| Mouse Expression Set 430 | 1454720_at | - | - |
| Mouse Expression Set 430 | 1454720_PM_at | - | - |
| Murine Genome U74v2 Set | 102750_at | - | - |
| Murine Genome U74v2 Set | 102750_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416182_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416182_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416182_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416182_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2510348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2510348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002510348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2660110 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2660110 | - | - |
| mouse-6 v1.1 (Illumina) | scl057267.10_22-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2510348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl057267.10_22-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2510348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2660110 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2660110 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl057267.10_22-S | - | - |

Apba3
Genomic Map