Assay Details
| Gene Symbol: Pfkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291071.1 | NP_001278000.1 | 9 - 10 | 1050 | 55 | |
| NM_019703.4 | NP_062677.1 | 9 - 10 | 1050 | 55 | ||
| GenBank mRNA | AF123533.1 | - | 9 - 10 | 1015 | 55 | |
| AK004790.1 | - | 9 - 10 | 1005 | 55 | ||
| AK049841.1 | - | 9 - 10 | 1004 | 55 | ||
| AK054193.1 | - | 9 - 10 | 1007 | 55 | ||
| AK076803.1 | - | 9 - 10 | 1005 | 55 | ||
| AK078254.1 | - | 9 - 10 | 1005 | 55 | ||
| AK098162.1 | - | 9 - 10 | 1004 | 55 | ||
| AK152670.1 | - | 9 - 10 | 1005 | 55 | ||
| AK154125.1 | - | 9 - 10 | 1005 | 55 | ||
| AK165422.1 | - | 9 - 10 | 1004 | 55 | ||
| AK165425.1 | - | 9 - 10 | 1004 | 55 | ||
| AK170624.1 | - | 9 - 10 | 1005 | 55 | ||
| AK171062.1 | - | 9 - 10 | 1005 | 55 | ||
| AY779275.1 | - | 9 - 10 | 967 | 55 | ||
| AY779276.1 | - | 9 - 10 | 967 | 55 | ||
| BC006926.1 | - | 9 - 10 | 989 | 2655383 | 55 | |
| CT010268.1 | - | 9 - 10 | 958 | 55 | ||
| Y19008.1 | - | 9 - 10 | 1007 | 55 | ||
More Information
Set Membership: |
|

Pfkp
Genomic Map