Assay Details
| Gene Symbol: Map2k2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023138.4 | NP_075627.2 | 8 - 9 | 1191 | 56 | |
| GenBank mRNA | AK009392.1 | - | 8 - 9 | 1208 | 56 | |
| AK011247.1 | - | 8 - 9 | 1185 | 56 | ||
| AK080574.1 | - | 8 - 9 | 1192 | 56 | ||
| AK171584.1 | - | 8 - 9 | 1180 | 56 | ||
| AK172055.1 | - | 8 - 9 | 1189 | 56 | ||
| AK181352.1 | - | 1 - 2 | 98 | 56 | ||
| AK186012.1 | - | 1 - 2 | 48 | 56 | ||
| AK208021.1 | - | 1 - 2 | 48 | 56 | ||
| AK217783.1 | - | 1 - 2 | 48 | 56 | ||
| BC014830.1 | - | 8 - 9 | 1170 | 4483473 | 56 | |
| S68267.1 | - | 8 - 9 | 1112 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k2
Genomic Map