Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000005432.mm.1 |
NM_001199271.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005432.mm.1 |
NM_001199271.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005432.mm.1 |
NM_031196.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005432.mm.1 |
NM_031196.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005432.mm.1 |
TC1000000752.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005433.mm.1 |
NM_001199271.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005433.mm.1 |
NM_001199271.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005433.mm.1 |
NM_031196.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005433.mm.1 |
NM_031196.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005433.mm.1 |
TC1000000752.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005431.mm.1 |
NM_001199271.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005431.mm.1 |
NM_001199271.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005431.mm.1 |
NM_031196.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005431.mm.1 |
NM_031196.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005431.mm.1 |
TC1000000752.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000005437.mm.1 |
NM_001199271.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005437.mm.1 |
NM_001199271.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005437.mm.1 |
NM_031196.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005437.mm.1 |
NM_031196.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000005437.mm.1 |
TC1000000752.mm.1 |
MTA TC |