Assay Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145146.2 | NP_660128.2 | 7 - 8 | 885 | 98 | |
| GenBank mRNA | AJ011080.1 | - | 7 - 8 | 98 | ||
| AK143987.1 | - | 7 - 8 | 886 | 98 | ||
| BC026681.1 | - | 7 - 8 | 803 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P425734 | BC100597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P425734 | AJ011080.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P425734 | NM_145146.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P425734 | AI255825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P425734 | BC100597.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P425734 | AJ011080.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P425734 | NM_145146.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P425734 | AI255825.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 78148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12948 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536060 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 839399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950531 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246659 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488520 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109317 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3031 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152323 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170420 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P425734 | BC100597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P425734 | AJ011080.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P425734 | NM_145146.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P425734 | AI255825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P425734 | BC100597.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P425734 | AJ011080.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P425734 | NM_145146.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P425734 | AI255825.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209597 | BC026681.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P209597 | AK143987.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883559 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23883534 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152323 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05651581 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170420 | - | - |
| Whole Mouse Genome G4122A | A_52_P425734 | BC100597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P425734 | AJ011080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P425734 | NM_145146.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P425734 | AI255825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P425734 | BC100597.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P425734 | AJ011080.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P425734 | NM_145146.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P425734 | AI255825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P209597 | BC026681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P209597 | AK143987.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137461 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152323 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170420 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05651586 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04882927 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04882924 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04882930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04882928 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04601801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23883557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04194219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04757426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04788831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04153941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04918315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04898113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04340739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883548 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04165243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04788839 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04181531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04781819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23883545 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10523102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523103 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523097 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523098 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523105 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523106 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523099 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523096 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523108 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523100 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10523101 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427561_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427440_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10523095 | - | - |
| Mouse Genome 430 2.0 Array | 1427561_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427440_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427440_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1427561_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 280662_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 280662_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500007363.mm.1 | NM_145146.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500007363.mm.1 | NM_145146.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500007363.mm.1 | TC0500000836.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427440_a_at | - | - |
| Mouse Expression Set 430 | 1427561_a_at | - | - |
| Mouse Expression Set 430 | 1427561_PM_a_at | - | - |
| Mouse Expression Set 430 | 1427440_PM_a_at | - | - |
| Murine Genome U74v2 Set | 94563_PM_at | - | - |
| Murine Genome U74v2 Set | 94563_at | - | - |
| Murine Genome U74v2 Set | 137170_PM_at | - | - |
| Murine Genome U74v2 Set | 137170_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427561_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427561_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427440_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427440_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427561_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427440_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427440_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427561_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 805103 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6770022 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006770022 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2678063 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2678063 | - | - |
| mouse-6 v1.1 (Illumina) | scl27606.14.1_65-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6770022 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27606.14.1_65-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770022 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2678063 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2678063 | - | - |

Afm
Genomic Map