Assay Details
| Gene Symbol: Terf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286200.1 | NP_001273129.1 | 9 - 10 | 1268 | 81 | |
| NM_009353.2 | NP_033379.2 | 8 - 9 | 1460 | 81 | ||
| NR_104410.1 | 9 - 10 | 1291 | 81 | |||
| GenBank mRNA | AF003000.1 | - | 8 - 9 | 1354 | 81 | |
| AK089419.1 | - | 9 - 10 | 1243 | 81 | ||
| AK160421.1 | - | 9 - 10 | 936 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Terf2
Genomic Map