Assay Details
| Gene Symbol: Mpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010822.3 | NP_034952.2 | 2 - 3 | 411 | 69 | |
| GenBank mRNA | AK018694.1 | - | 2 - 3 | 391 | 69 | |
| AK076201.1 | - | 2 - 3 | 406 | 69 | ||
| AK088878.1 | - | 2 - 3 | 365 | 69 | ||
| AK156067.1 | - | 2 - 3 | 378 | 69 | ||
| BC014754.1 | - | 2 - 3 | 405 | 4486938 | 69 | |
| U10420.1 | - | 2 - 3 | 404 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpg
Genomic Map